S.O.C medium, Corning®
Catalog # MDTC46-003-CR
Supplier: Corning
CAS Number:
Specifications
- Pk:10
- Description:S.O.C medium
- Environmentally Preferable:
Specifications
About this item
S.O.C
The use of SOC medium improves transformation efficiency.
medium is used as a recovery medium during the final stage of transformation of E. coli.