Captiva EMR-Lipid cartridges
Supplier: Agilent
Certificates
About this item
Captiva EMR–Lipid provides highly selective and efficient lipid/matrix removal without unwanted analyte loss.
- Highly selective and efficient lipid removal: The unique EMR Lipid mechanism combines size exclusion and hydrophobic interactions between the sorbent and the long aliphatic chain of lipids
- Clog-free operation: Advanced filter design and construction technology ensures an easy flow
- Time savings and improved precision: A protein crash solvent retention frit in 1 ml and 96-well plate formats promotes a streamlined, automatable in-well protein precipitation workflow
The novel EMR-Lipid technology removes lipids based on a combination of size exclusion and hydrophobic interaction. Effective lipid removal assures minimal ion suppression of target analytes, which significantly improves method reliability and ruggedness. The 96-well plate and 1 ml cartridge formats contain a solvent retention frit, which allows solvent-first in-well protein precipitation which streamlines sample preparation. The improved filter design gives easy flow with vacuum or positive pressure. The 3 and 6 ml cartridge formats allow gravity flow with the absence of solvent retention frits and are easy to use.
Frequently Bought Together