0 zł
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Poland
ContactUsLinkComponent
5-Bromo-1-methyl-1H-1,2,4-triazole
5-Bromo-1-methyl-1H-1,2,4-triazole 97%
5-Bromo-1-methyl-1H-1,2,4-triazole 97%
APOSOR904770-5G
:
Apollo Scientific
:
5-Bromo-1-methyl-1H-1,2,4-triazole 97%
APOSOR904770-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
Environmentally Preferable:
:
:
161.9900 g/mol
:
Ambient
:
16681-72-4
:
C₃H₄BrN₃
:
MFCD11109756