Silver bromide
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Argentous bromide
CAS Number:7785-23-1
:
Formula:AgBr
MDL Number:MFCD00003398
Boiling Pt:1300
Molecular Weight:187.77 g/mol
Melting Pt:432 °C
ADR:9+EHS III
Merck Index:12,08649
UN:3077
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together