3-(4-Pyridinyl)-2-propyn-1-ol 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-(4-Pyridyl)-2-propyn-1-ol
CAS Number:93524-95-9
:
Formula:C₈H₇NO
MDL Number:MFCD00168868
Molecular Weight:133.147 g/mol
Melting Pt:117…119 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together