Order Entry
Poland
ContactUsLinkComponent
6-Chloro-1H-indole-2-carbaldehyde 95%
6-Chloro-1H-indole-2-carbaldehyde 95%
Catalog # APOSOR927744-1G
Supplier:  Apollo Scientific
CAS Number:  
6-Chloro-1H-indole-2-carbaldehyde 95%
Catalog # APOSOR927744-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047
  • Environmentally Preferable:

Specifications

About this item

 :
 :179.603 g/mol
Storage Temperature:Ambient
CAS Number:53590-59-3
Formula:C₉H₆ClNO
MDL Number:MFCD11040767