Flame resistant anti-static fleece jackets, Modaflame™, FR30
: Portwest
These Modaflame™ Fleece Jackets are constructed with a double-sided anti-pill fleece. Features include two side pockets with concealed zip closure to keep belongings secure. Comfortable elasticated cuffs and drawstring at hem.
- Drawcord adjustable hem
- Front zip opening
- Zipped handwarmer pockets
- 50+ UPF rated fabric to block 98% of UV rays
- Shell fabric is made of Modaflame™ Knit (280 g)
These Modaflame™ garments can be layered to achieve an overall Arc Thermal Performance Value (ATPV) or Energy Break open threshold (Ebt) Rating. These industry leading range provides multi standard protection for hazardous environments.
: EN ISO 11612 (A1, B1, C2); EN 1149.
- Catalog No:
- PRTWFR30NARM
- PRTWFR30NARL
- PRTWFR30NARXXXL
- PRTWFR30NARXL
- PRTWFR30NARXXL
- PRTWFR30NARS
- Model:
- FR30
- FR30
- FR30
- FR30
- FR30
- FR30
- Garment application:
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Fire protection
- Risk area:
- Heat, Flame-retardant
- Heat, Flame-retardant
- Heat, Flame-retardant
- Heat, Flame-retardant
- Heat, Flame-retardant
- Heat, Flame-retardant
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Zip closure
- Fabric:
- Modaflame ™
- Modaflame ™
- Modaflame ™
- Modaflame ™
- Modaflame ™
- Modaflame ™
- Weight:
- 280 g
- 280 g
- 280 g
- 280 g
- 280 g
- 280 g
Frequently Bought Together