Iron(II) fluoride, anhydrous 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ferrous fluoride, Iron difluoride
CAS Number:7789-28-8
:
Formula:F₂Fe
MDL Number:MFCD00016093
Boiling Pt:1100
Melting Pt:>1000 °C (subl.)
Density:4.09
Molecular Weight:93.84 g/mol
EINECS:232-155-7
ADR:8 II
Merck Index:14,4045
UN:3260
:GHS05
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together