Lithium perchlorate, Sigma-Aldrich®
: Merck
:7791-03-9
:
:ClLiO₄
:MFCD00011079
:430
:236…237 °C
:2.42 g/cm³ (20 °C)
:106.39 g/mol
:232-237-2
:1481
:5.1 II
:13,05561
:M-Clarity™ quality level = MQ100
:GHS03
:GHS07
:Ambient
- Environmentally Preferable:
Frequently Bought Together