5,5'-Dithiobis(2-nitrobenzoic acid) (Ellmans reagent, DTNB), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: DTNB, Ellman's reagent, 3,3'-Dithiobis(6-nitrobenzoic acid), 5,5'-Disulphanediylbis(2-nitrobenzoic acid), 2,2'-Dinitro-5,5'-dithiodibenzoic acid
CAS Number:69-78-3
:
Formula:C₁₄H₈N₂O₈S₂
MDL Number:MFCD00007140
Molecular Weight:396.36 g/mol
Melting Pt:240…244 °C
:M-Clarity™ quality level = MQ200
EINECS:200-714-4
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together