Itaconic acid, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 2-Methylenesuccinic acid, Methylenesuccinic acid
CAS Number:97-65-4
:
Formula:C₅H₆O₄
MDL Number:MFCD00004260
Boiling Pt:381
Melting Pt:162…167 °C
Density:1.573 g/cm³ (20 °C)
Molecular Weight:130.1 g/mol
Merck Index:13,05264
:M-Clarity™ quality level = MQ200
EINECS:202-599-6
:GHS05
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together