Heptanoyl chloride ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Heptanoylchloride
CAS Number:2528-61-2
:
Formula:C₇H₁₃ClO
MDL Number:MFCD00000763
Boiling Pt:172…174 °C
Melting Pt:–84 °C
Density:0.960
Molecular Weight:148.63 g/mol
Flash Pt:58 °C (136 °F)
Storage Temperature:Ambient
EINECS:219-775-3
UN:2920
ADR:8 II
:GHS05
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together