Petrochemical filter stick apparatus with hooks and springs
Catalog # SCERSP513880-0000
Supplier: DWK Life Sciences
Specifications
- Description:Petrochemical filter stick apparatus
- Environmentally Preferable:
- Pk:1
Specifications
About this item
The Filter Stick Apparatus is used in the determination of oil content and solvent extractables in petroleum waxes.
- Supplied with one pair of springs
- Designed from ASTM D721 specifications
- Manufactured from borosilicate 3,3 glass
Assembly consists of a sintered glass filter stick with air pressure inlet tube and delivery nozzle and a cooling tube with NS 24/40 joint.
Certifications: The borosilicate glass conforms to USP Type I and ASTM E438, Type I, Class A requirements.