Europium triacetate hydrate ≥99.9% (REO, rare earth oxide basis), REacton®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Acetic acid europium (III) salt hydrate, Europium(III) acetate xhydrate, Europium (III) acetate hydrate
CAS Number:62667-64-5
:
Formula:C₆H₉EuO₆·H₂O
MDL Number:MFCD00150115
Molecular Weight:347.11 g/mol
Storage Temperature:Ambient
EINECS:214-670-9
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together