DEC-AHOL WFI® , DEC-AHOL AEROSOL WFI Formula, 70% Isopropyl alcohol and 30% WFI for cleanroom use
Supplier: VELTEK ASSOCIATES
New Product
Certificates
About this item
DEC-AHOL WFI® Formula is a 70% USP Isopropyl Alcohol (IPA) formulated with 30% Water for Injection (WFI). This USP IPA has been designed specifically for pharmaceutical, biotechnology, healthcare, and medical device cleaning rotations for use in both aseptic and non-aseptic environments. DEC-AHOL WFI Formula can be used for the decontamination of numerous surfaces within any cleanroom operation.
- Filled in an ISO 5 (Grade A/B, Former Class 100)
- Filtered at 0.2 microns
- Gamma irradiated at a 10-6 SAL
- Lot sterility tested according to current USP compendium
- Completely lot traceable from start to finish
- Completely validated for sterility and shelf life
Applications: In spray and wipe downs on hard, non-porous, inanimate surfaces; aseptic filling, gowning rooms, general manufacturing areas, process lines, machinery, tools, tables, counters, laminar flow benches, carts, shelves. Also for exterior packaging, accessories, glass, plastic, vinyl, stainless steel and cleaning gloves.
Certifications: DEC-AHOL WFI Formula is an EPA registered hard surface disinfectant and sanitizer when used as directed. Each lot of DEC-AHOL WFI Formula is sterility tested according to current USP Compendium, is completely traceable, and has been completely validated for sterility and shelf life. All shipments of DEC-AHOL WFI Formula are delivered with lot specific certificates.
Frequently Bought Together