Specifications
- Description:Powered air purifying respirator starter kit, Versaflo™ TR-819E IS
- Seq:01
- Environmentally Preferable:
- Pk:1
Specifications
About this item
The TR-819E starter kit for the Intrinsically Safe (IS) powered air purifying respirator comes ready to use and includes everything needed for immediate use.
- High level of respiratory protection
- Compatible with filters for TR-600
- Includes a rechargeable lithium-ion battery with standard capacity
- Three selectable airflow rates for high comfort
- Blower unit with intuitive, bright yellow touch points
- Audible, visual and vibratory alarms for low airflow and low battery
- Ergonomic weight distribution ensures high comfort to the wearer
The ATEX TR 800 certified powered air purifying respirator from the Versaflo™ product family provides long lasting protection against particles, gases and vapours when working under difficult conditions, and in potentially explosive atmospheres is comfortable to wear at the same time.
Applicable in areas like chemical industry, food industry, pharmaceutical industry, metal processing, medicine and healthcare, paper industry, coking plants and foundries, paint spraying/coating and construction industry/building renovation.
The kit is approved to zone zero requirements and so may be suitable in areas where potentially explosive atmospheres are expected and permanent within normal operations.