Order Entry
Poland
ContactUsLinkComponent
 
Glycerine 98.0-101.0% (by alkalimetry) Reag. Ph. Eur., ACS
 :  Merck
    
 
undefined
Glycerine 98.0-101.0% (by alkalimetry) Reag. Ph. Eur., ACS
 :  Merck
    
 

 

  • Environmentally Preferable:

 

 

  • Assay (GC, Calculated on Anhydrous Substance):
    ≤99.5 %
  • Assay (Alkalimetric, Calculated on Anhydrous Substance):
    98.0 - 101.0%
  • Identity:
    Passes Test
  • Colour:
    ≤10 APHA
  • Acidity or Alkalinity:
    Passes Test
  • Free Acid (CH₃COOH):
    ≤0.002 %
  • Neutrality:
    Passes Test
  • Refractive Index (n 20/D):
    1.470 - 1.475
  • Chloride (Cl):
    ≤0.0010%
  • Sulphate (SO₄):
    ≤0.001%
  • Halogenated Compounds (as Cl):
    ≤0.0030%
  • Heavy Metals (as Pb):
    ≤0.00020%
  • Al (Aluminium):
    ≤0.00005%
  • B (Boron):
    ≤0.000002%
  • Ba (Barium):
    ≤0.00001%
  • Ca (Calcium):
    ≤0.0001%
  • Cd (Cadmium):
    ≤0.000005%
  • Co (Cobalt):
    ≤0.000005%
  • Cr (Chromium):
    ≤0.000002%
  • Cu (Copper):
    ≤0.000005%
  • Fe (Iron):
    ≤0.00001%
  • Mg (Magnesium):
    ≤0.0001%
  • Mn (Manganese):
    ≤0.00001%
  • Ni (Nickel):
    ≤0.000002%
  • Pb (Lead):
    ≤0.00001%
  • Sn (Tin):
    ≤0.00001%
  • Zn (Zinc):
    ≤0.00001%
  • Methanol:
    ≤3000 ppm
  • Other Residual Solvents (ICH Q3C):
    Excluded by Production Process
  • Acrolein and Glucose:
    Passes Test
  • Aldehydes:
    ≤10 ppm
  • Diethylene Glycol and Related Substances (Impurity A):
    ≤0.1%
  • Diethylene Glycol and Related Substances (Any Further Impurity):
    ≤0.1%
  • Diethylene Glycol and Related Substances (Sum of All Impurities):
    ≤0.5%
  • Fatty Acids and Esters (as Butyric Acid):
    ≤0.05%
  • Substances Darkened by Sulphuric Acid:
    Passes Test
  • Sugar:
    Passes Test
  • Sulfated Ash:
    ≤0.005%
  • Water:
    ≤0.5%

 

Frequently Bought Together