Arsenic(III) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: di-Arsenic trioxide
CAS Number:1327-53-3
:
Formula:As₂O₃
MDL Number:MFCD00003433
Boiling Pt:465
Melting Pt:312
Density:3.8 g/cm³ (20 °C)
Molecular Weight:197.84 g/mol
Storage Temperature:Ambient
EINECS:215-481-4
UN:1561
ADR:6.1 II
Merck Index:12,00844
:M-Clarity™ quality level = MQ200
:GHS06
:GHS05
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together