Calcium hydrogen orthophosphate, anhydrous 97%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Calcium phosphate dibasic, Calcium hydrogen phosphate, Calcium perphosphate, ammonified, Anhydrous Dibasic Calcium Phosphate, Calcium phosphate, dibasic
CAS Number:7757-93-9
:
Formula:CaHPO₄
MDL Number:MFCD00010909
Molecular Weight:136.06 g/mol
Melting Pt:370
Merck Index:13,01697
Storage Temperature:Ambient
EINECS:231-826-1
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White Powder
- Titration Complexometric:≥96.0 %
Specification Test Results
Frequently Bought Together