Order Entry
Poland
ContactUsLinkComponent
Magnesium perchlorate ACS desiccant
Magnesium perchlorate ACS desiccant
Certificates
undefined
Magnesium perchlorate ACS desiccant
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    White Crystalline powder or flakes and/or chunks
  • Loss on drying:
    ≤8 % (1 g, 190°C)
  • Suitability:
    (for moisture absorption) Passes test
  • Titrable base:
    ≤0.025 meq/g
  • Titrable free acid:
    ≤0.005 meq/g