Triphenylamine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:603-34-9
:
Formula:C₁₈H₁₅N
MDL Number:MFCD00003020
Boiling Pt:347
Melting Pt:123…128 °C
Density:0.774 g/cm³ (20 °C)
Molecular Weight:245.32 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:210-035-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together