Methyl-4-nitrobenzoate, Sigma-Aldrich®
Catalog # SIAL72962-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-Nitrobenzoic acid emthyl ester, Methyl 4-nitrobenzoate
:
:181.15 g/mol
Melting Pt:94…96 °C
Storage Temperature:Ambient
EINECS:210-599-2
CAS Number:619-50-1
Formula:C₈H₇NO₄
MDL Number:MFCD00007350
:M-Clarity™ quality level = MQ200