1-Undecanol, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane
- CAS Number:112-42-5
- Formula:C₁₁H₂₄O
- MDL Number:MFCD00004751
- Molecular Weight:172.31 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:13…15 °C
- Density:0.83 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:203-970-5
- UN:3082
- ADR:9,III
- :M-Clarity™ quality level = MQ300
2 Options Available Below
Product Details & Documents
Synonyms: 1-Undecyl alcohol, Undecyl alcohol, Undecan-1-ol, Decyl carbinol, 1-Hydroxy undecane
- CAS Number:112-42-5
- Formula:C₁₁H₂₄O
- MDL Number:MFCD00004751
- Molecular Weight:172.31 g/mol
- Boiling Pt:243 °C (1013 hPa)
- Melting Pt:13…15 °C
- Density:0.83 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:203-970-5
- UN:3082
- ADR:9,III
- :M-Clarity™ quality level = MQ300
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



