Ethyl propionate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Propionic acid ethyl ester
CAS Number:105-37-3
:
Formula:C₅H₁₀O₂
MDL Number:MFCD00009308
Boiling Pt:210
Melting Pt:–74 °C
Density:0.89 g/cm³ (20 °C)
Molecular Weight:102.13 g/mol
Flash Pt:12
Storage Temperature:Ambient
EINECS:203-291-4
UN:1195
ADR:3 II
Merck Index:13,03878
:M-Clarity™ quality level = MQ100
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together