Order Entry
Poland
ContactUsLinkComponent
Hexanoyl chloride 97%
Hexanoyl chloride 97%
Certificates
undefined
Hexanoyl chloride 97%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4250 to 1.4270 (20°C, 589 nm)
  • GC:
    ≥96.5 %

Specification Test Results

Related Information

Frequently Bought Together