L(-)-Ephedrine
Catalog # PROONMIAM723
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: (1R,2S)-(-)-Ephedrine
:
:165.24
EINECS:206-080-5
UN:1544
:GHS07
CAS Number:299-42-3
Formula:C₆H₅CH(OH)CH(CH₃)NHCH₃
Boiling Pt:132 °C (3 mmHg)
Melting Pt:217…218 °C
Density:1.124 g/cm³ (20 °C)
ADR:6.1 III