Scandium(III) chloride hexahydrate, Sigma-Aldrich®
Catalog # SIAL451274-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:259.41 g/mol
EINECS:233-799-1
CAS Number:20662-14-0
Formula:Cl₃Sc·6H₂O
MDL Number:MFCD00149852
:M-Clarity™ quality level = MQ100