tetra-Chloroauric(III) acid hydrate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Gold(III) chloride hydrate, Hydrogen tetrachloroaurate(III) xhydrate, Hydrogen tetrachloroaurate (III) hydrate, Hydrogen tetrachloroaurate hydrate, Chloroauric acid, tetra-Chloroauric acid hydrate
CAS Number:27988-77-8
:
Formula:HAuCl₄·H₂O
MDL Number:MFCD00149903
Density:1.169 g/cm³ (20 °C)
Molecular Weight:357.8 g/mol
ADR:8 III
:M-Clarity™ quality level = MQ100
EINECS:240-948-4
UN:3260
:GHS05
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together