(±)-1-Phenyl-1-propanol, Sigma-Aldrich®
SIAL111309-50G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:50 g
- Seq:3368
- Environmentally Preferable:
: 1-Phenyl-1-propanol, (±)-α-Ethylbenzyl alcohol
:
:136.19 g/mol
:∼219 °C (1013 hPa)
:202-256-0
:GHS07
:99
:Ambient
:93-54-9
:C₆H₅CH(OH)CH₂CH₃
:MFCD00004564
:0.99 g/cm³ (20 °C)
:13,03804
:M-Clarity™ quality level = MQ100