Tween® 80 (Polysorbate), Sigma-Aldrich®
: Merck
:9005-65-6
:
:MFCD00082107
:∼100 °C (1013 hPa)
:–21 °C
:1.062 g/cm³ (20 °C)
:>149 °C
:13,07664
:M-Clarity™ quality level = MQ200
:Ambient
:500-019-9
- Environmentally Preferable:
Frequently Bought Together