Order Entry
Poland
ContactUsLinkComponent
Poly(methacrylic acid, sodium salt) solution, M.W. 4000 - 6000 40% (w/w) in water, Sigma-Aldrich®
Poly(methacrylic acid, sodium salt) solution, M.W. 4000 - 6000 40% (w/w) in water, Sigma-Aldrich®
Catalog # SIAL674044-1L
Supplier:  Merck
CAS Number:  
Poly(methacrylic acid, sodium salt) solution, M.W. 4000 - 6000 40% (w/w) in water, Sigma-Aldrich®
Catalog # SIAL674044-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Infrared spectrum:
    Conforms to structure
  • Solid content:
    38.0 - 42.0%
  • Concentration:
    40 wt.% in H₂O
  • Refractive index:
    n20/D 1.430

Specification Test Results

About this item

Synonyms: Sodium polymethacrylate, Sodium methacrylate polymer, Methacrylic acid, sodium salt
 :
 :108.071 g/mol
CAS Number:54193-36-1
Formula:C₄H₅NaO₂
MDL Number:MFCD00156535
Density:1.251 g/ml (25 °C)
 :M-Clarity™ quality level = MQ100