D-(-)-Arabinose ≥99%, powder
Supplier: MP Biomedicals
Certificates
About this item
Synonyms: D-(-)-Arabinose, (2S,3R,4R)-2,3,4,5-Tetrahydroxypentanal
CAS Number:10323-20-3
:28697-53-2
Formula:C₅H₁₀O₅
MDL Number:MFCD00135608
Molecular Weight:150.13 g/mol
Melting Pt:160
Storage Temperature:Ambient
EINECS:233-708-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together