D-(+)-Xylose
Catalog # MOLE12518844-500G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
About this item
:
:150.13 g/mol
Boiling Pt:331
Melting Pt:154
Density:1.525 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-400-7
CAS Number:58-86-6
Formula:C₅H₁₀O₅
MDL Number:MFCD00151475
Merck Index:13,10142