Paraffin, Solidification point: 58-60°C, granules, TECHNICAL
Supplier: VWR Chemicals
Certificates
About this item
Synonyms: Wax Paraffin, Paraffin wax
CAS Number:8002-74-2
:64742-51-4
Melting Pt:58…60 °C
Density:0.9 g/cm³ (20 °C)
EINECS:232-315-6
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Identification:Passes test
Specification Test Results
Related Information
Certificates
Search by Lot Number2,5L Plastic bottle for solids DIN 80 (ZPACK13252500)
Packaging Information
27511289.pdf spec_VWRC26154.291 spec_VWRC26154.460 spec_VWRC26154.460
Product specification
Frequently Bought Together