Nickel ≥99.98% (metals basis), wire, annealed, Ø 1.0 mm (0.04 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-02-0
:
Formula:Ni
MDL Number:MFCD00011137
Boiling Pt:3007
Melting Pt:1453
Density:8.9 g/cm³ (20 °C)
Molecular Weight:58.69 g/mol
Storage Temperature:Ambient
EINECS:231-111-4
ADR:4.1 III
Merck Index:12,06582
UN:3089
:GHS08
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together