Titanium ≥99.7% (metals basis), foil, annealed, Thickness: 0.032 mm (0.0013 in)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7440-32-6
:
Formula:Ti
MDL Number:MFCD00011264
Boiling Pt:3287 °C
Melting Pt:1660
Density:4.50
Molecular Weight:47.88 g/mol
Merck Index:12,09610
EINECS:231-142-3
:GHS02
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together