Anti-Estrogen Receptor Polyclonal Antibody
Catalog # BSBTPB9192
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen symbol:Estrogen Receptor
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Polyclonal antibody for ESTROGEN RECEPTOR ALPHA/ESR1 detection. Host: Rabbit.Size: 100μg/vial. Tested applications: IHC-P. Reactive species: Human. ESTROGEN RECEPTOR ALPHA/ESR1 information: Molecular Weight: 66216 MW; Subcellular Localization: Isoform 1: Nucleus . Cytoplasm . Cell membrane ; Peripheral membrane protein ; Cytoplasmic side . A minor fraction is associated with the inner membrane; Tissue Specificity: Widely expressed. Isoform 3 is not expressed in the pituitary gland.
Type: Primary
Antigen: Estrogen Receptor
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype: Rabbit
Reactivity: Human