Human recombinant PLGF 1 (from E. coli)
PRSI40-529
:
- Pk:25 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Environmentally Preferable:
- Biological activity:Biological activity was determined by its ability to chemoattract human monocytes of using a concentration range of 1.0-10.0 ng/mL.
- Gene ID:5228
- Protein synonyms:Placenta growth factor|D12S1900|PLGF|PGFL|PlGF|PlGF-2|SHGC-10760
- Protein/peptide name:PLGF 1
- Purity:>95%
- Formulation:Lyophilized
PlGF-1 is an angiogenic factor that belongs to the cysteine-knot superfamily of growth factors. PlGF-1 is expressed in placental tissues, colon and mammary carcinomas. It signals through the VEGFR-1/FLT1 receptor and stimulates endothelial cell proliferation and migration. Recombinant human PlGF-1 is a 29.7 kDa disulfide-linked homodimeric protein of two 131 amino acid polypeptide chains.
- Store in working aliquots at -20°C. Avoid freeze/thaw cycles.