Anti-SYNPR Rabbit Polyclonal Antibody
Catalog # PRSI50-192
Supplier: ProSci Inc.
Specifications
- Antigen name:Synaptoporin
- Antigen symbol:SYNPR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Antigen synonyms:SPO
- Immunogen:Peptide corresponding to amino acid residues from the C-terminal region of rat synaptophysin 2, conjugated to keyhole limpet hemocyanin (KLH).
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Synaptophysin 2, also known as synaptoporin, is a component of the synaptic vesicle membrane and thought to play an important role in synaptic vesicle trafficking (Dai et al., 2003). It is highly homologous to synaptophysin 1, yet differs in expression. Whereas synaptophysin 1 is ubiquitiously expressed, synaptophysin 2 is concentrated in the mossy fiber synapses of the hippocampus (Grabs et al., 1994; Singec et al., 2002).
Affinity Purified
Affinity Purified
100 uL in 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per mL BSA and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SYNPR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat