Order Entry
Poland
ContactUsLinkComponent
Anti-AGAP2 Goat Polyclonal Antibody
Anti-AGAP2 Goat Polyclonal Antibody
Catalog # PRSI46-997
Supplier:  ProSci Inc.
undefined
Anti-AGAP2 Goat Polyclonal Antibody
Catalog # PRSI46-997
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    ArfGAP With GTPase Domain, Ankyrin Repeat And PH Domain 2
  • Antigen symbol:
    AGAP2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    GGAP2|CENTG1|PIKE
  • Concentration:
    500 ug/mL
  • Immunogen:
    PIKE antibody was raised against a 15 amino acid synthetic peptide near the internal region of PIKE.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

PIKE antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

PIKE antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AGAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human