Order Entry
Poland
ContactUsLinkComponent
Anti-F13A1 Goat Polyclonal Antibody
Anti-F13A1 Goat Polyclonal Antibody
Catalog # PRSI45-564
Supplier:  ProSci Inc.
undefined
Anti-F13A1 Goat Polyclonal Antibody
Catalog # PRSI45-564
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Coagulation Factor 13 A1
  • Antigen symbol:
    F13A1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Antigen synonyms:
    coagulation factor XIII A chain|A1 polypeptide)|protein-glutamine gamma-glutamyltransferase A chain|A subunit|coagulation factor XIII|coagulation factor XIIIa|fibrinoligase|FSF|bA525O21.1 (coagulation factor XIII|factor XIIIa|TGase|A polypeptide|fibrin stabilizing factor|transglutaminase. plasma|transglutaminase A chain
  • Concentration:
    500 ug/mL
  • Immunogen:
    Factor XIII alpha antibody was raised against a 15 amino acid synthetic peptide near the internal region of Factor XIII alpha (aa 703-717).
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Factor XIII alpha antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.

Factor XIII alpH a antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.

Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: F13A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human