Anti-GLYCTK Rabbit Polyclonal Antibody
PRSI30-315
- Antigen name:Glycerate Kinase
- Antigen symbol:GLYCTK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:HBEBP4|HBeAgBP4A|HBEBP2
- Immunogen:A synthetic peptide corresponding a region of human GLYCTK.
- Size:100 µg
- Pk:100 µG
Deficiency of human glycerate kinase leads to D-glycerate acidemia/D-glyceric aciduria.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GLYCTK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog