Anti-GNB2L1 Rabbit Polyclonal Antibody
PRSI29-362
- Antigen name:Guanine Nucleotide Binding Protein (G Protein), Beta Polypeptide 2-like 1
- Antigen symbol:GNB2L1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Zebrafish,Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:H12.3|Receptor for activated C kinase|P205|Gnb2-rs1|Receptor of activated protein kinase C 1|PIG21|Guanine nucleotide-binding protein beta subunit-like protein|HLC-7|RACK1
- Immunogen:A synthetic peptide corresponding a region of human GNB2L1.
- Size:50 µg
- Pk:50 µG
GNB2L1 seems to bind protein kinase C acting as an intracellular receptor to anchor the activated PKC to the cytoskeleton.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: GNB2L1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Dog, Zebrafish