- Antigen name:Dimethylarginine dimethylaminohydrolase 2
- Antigen symbol:DDAH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:DDAHII|NG-dimethylarginine dimethylamino hydrolase homolog|DDAH2|G6a|OTTHUMP00000029307|NG30|OTTHUMP00000174406|dimethylarginine dimethylaminohydrolase II|OTTHUMP00000062666|DADB-110M10.5|DDAH|alternat|dimethylarginine dimethylaminohydrolase 2
- Immunogen:A synthetic peptide corresponding a region of human DDAH2.
- Size:50 µg
- Pk:50 µG
DDAH2 hydrolyzes N (G),N (G)-dimethyl-L-arginine (ADMA) and N (G)-monomethyl-L-arginine (MMA) which act as inhibitors of NOS. DDAH2 has therefore a role in nitric oxide generation.This gene belongs to the dimethylarginine dimethylaminohydrolase (DDAH) gene family. The encoded enzyme plays a role in nitric oxide generation by regulating cellular concentrations of methylarginines, which in turn inhibit nitric oxide synthase activity. Publication Note: This RefSeq record includes a subset of the publications that are available for this gene. Please see the Entrez Gene record to access additional publications.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: DDAH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat