Specifications
- Antigen name:Splicing Factor, Suppressor Of White-Apricot Family
- Antigen symbol:SFSWAP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:Lyophilized
- Antigen synonyms:SWAP|SFRS8
- Immunogen:A synthetic peptide corresponding a region of human SFRS8.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
SFRS8 is a human homolog of Drosophila splicing regulatory protein.This gene encodes a human homolog of Drosophila splicing regulatory protein. This gene autoregulates its expression by control of splicing of its first two introns. In addition, it also regulates the splicing of fibronectin and CD45 genes. Multiple alternatively spliced variants have been identified although their full-length natures have not been characterized to date.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SFSWAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse