Order Entry
Poland
ContactUsLinkComponent
Nitric acid 2 M HNO3 (2N) volumetric solution, Fluka™
Nitric acid 2 M HNO3 (2N) volumetric solution, Fluka™
Catalog # 35278-1L
CAS Number:  
Nitric acid 2 M HNO3 (2N) volumetric solution, Fluka™
Catalog # 35278-1L
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Factor:
    0.999 - 1.001
  • BAM/EMPA reference material:
    effective
  • BAM/EMPA Certificate Number:
    effective
  • Traceable to NIST SRM:
    effective

Specification Test Results

About this item

Synonyms: Nitric acid solution
 :
 :63.01 g/mol
Boiling Pt:100 °C
UN:2031
 :GHS05
Melting Pt:-42
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349
Density:1.065 g/cm³ (20 °C)
ADR:8 II
Merck Index:13,06610