Anti-EPGN Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-5767R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:epithelial mitogen
- Antigen symbol:EPGN
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:255324
- Antigen synonyms:ALGV3072|PRO9904|EPG
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Epigen is an EGF-related polypeptide growth factor that signals through the ErbB receptor-1. It is produced in several tissues, including the testis, liver, heart and in certain tumor cells. Epigen is mitogenic for fibroblasts and epithelial cells. Human Epigen is initially synthesized as a glycosylated 14.7 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a mature soluble sequence.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: EPGN
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat