Anti-MDH1 Rabbit Polyclonal Antibody (Cy7®)
Catalog # BOSSBS-3996R-CY7
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Malate Dehydrogenase 1
- Antigen symbol:MDH1
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:4190
- Antigen synonyms:MOR2|MDH-s|MGC:1375|cytoplasmic|HEL-S-32|Malate dehydrogenase|EC=1.1.1.37|Cytosolic malate dehydrogenase|MDHA
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:284-334/334
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Malate dehydrogenase catalyzes the reversible oxidation of malate to oxaloacetate, utilizing the NAD/NADH cofactor system in the citric acid cycle. Malate dehydrogenase 1 (MDH1) is localized to the cytoplasm and may play pivotal roles in the malate-aspartate shuttle that operates in the metabolic coordination between cytosol and mitochondria.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: MDH1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat