Anti-MTNR1A Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-0027R-CY5
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Melatonin Receptor 1A
- Antigen symbol:MTNR1A
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Bovine,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:4543
- Antigen synonyms:MEL-1A-R|MEL1A|melatonin receptor type 1A|melatonin 1|melatonin 1A|MT1|mel1a receptor
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:209-242/350
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
High affinity receptor for melatonin. Likely to mediates the reproductive and circadian actions of melatonin. The activity of this receptor is mediated by pertussis toxin sensitive G proteins that inhibit adenylate cyclase activity.
Recommended Dilutions: IF(IHC-P): 1:50-200; IF(ICC)(1:50-200
Type: Primary
Antigen: MTNR1A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine