Anti-CRF Rabbit Polyclonal Antibody [clone: R153]
Catalog # BWRLBS2869
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CRF
- Clonality:Polyclonal
- Clone:R153
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 121-170 of Human CRF.
CRF (R153) pAb detects endogenous levels of CRF protein.
Type: Primary
Antigen: CRF
Clonality: Polyclonal
Clone: R153
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat